Assay Details
| Gene Symbol: Taf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177342.3 | NP_796316.2 | 5 - 6 | 1438 | 97 | |
| GenBank mRNA | AK031985.1 | - | 5 - 6 | 1442 | 97 | |
| AK088953.1 | - | 5 - 6 | 1438 | 97 | ||
| AK169453.1 | - | 5 - 6 | 1438 | 97 | ||
| BU057827.1 | - | 3 - 4 | 536 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf5
Genomic Map