Assay Details
| Gene Symbol: Cdkn2aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078329.1 | - | 2 - 3 | 504 | 108 | |
| AK089057.1 | - | 2 - 3 | 507 | 108 | ||
| AK167684.1 | - | 2 - 3 | 505 | 108 | ||
More Information
Set Membership: |
|

Cdkn2aip
Genomic Map