Assay Details
| Gene Symbol: A830011I04 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043597.1 | - | 1 - 2 | 89 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
bromodomain, testis-specific |
Brdt |
7420412D09Rik, Brd6, Fsrg3 |
-42.7kb | Anti-sense | Chr. 5 - 107331091 - 107387061 on NCBI Build GRCm38 |
| Downstream | BTB (POZ) domain containing 8 | Btbd8 |
EG627196, Gm16115, OTTMUSG00000030610 |
6.1kb | Anti-sense | Chr. 5 - 107437997 - 107491596 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

A830011I04
Genomic Map