| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900004715.mm.1 |
NM_001164336.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004715.mm.1 |
NM_001164336.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004715.mm.1 |
NM_001164337.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004715.mm.1 |
NM_001164337.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004715.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004715.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004715.mm.1 |
TC1900000527.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004719.mm.1 |
NM_001164336.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004719.mm.1 |
NM_001164336.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004719.mm.1 |
NM_001164337.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004719.mm.1 |
NM_001164337.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004719.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004719.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004719.mm.1 |
TC1900000527.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004720.mm.1 |
NM_001164336.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004720.mm.1 |
NM_001164336.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004720.mm.1 |
NM_001164337.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004720.mm.1 |
NM_001164337.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004720.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004720.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004720.mm.1 |
TC1900000527.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004717.mm.1 |
NM_001164336.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004717.mm.1 |
NM_001164336.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004717.mm.1 |
NM_001164337.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004717.mm.1 |
NM_001164337.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004717.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004717.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004717.mm.1 |
TC1900000527.mm.1 |
MTA TC |