Assay Details
| Gene Symbol: March5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164336.1 | NP_001157808.1 | 4 - 5 | 875 | 90 | |
| NM_027314.3 | NP_081590.3 | 4 - 5 | 875 | 90 | ||
| GenBank mRNA | AK007511.1 | - | 4 - 5 | 711 | 90 | |
| AK009232.1 | - | 4 - 5 | 861 | 90 | ||
| AK178944.1 | - | 3 - 4 | 90 | |||
| AK206577.1 | - | 2 - 3 | 270 | 90 | ||
| BC064752.1 | - | 4 - 5 | 599 | 30049608 | 90 | |
| BC107214.1 | - | 4 - 5 | 582 | 90 | ||
| BC107215.1 | - | 4 - 5 | 587 | 40051628 | 90 | |
| BC132455.1 | - | 4 - 5 | 639 | 40130732 | 90 | |
| BC132457.1 | - | 4 - 5 | 638 | 40130734 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

March5
Genomic Map