| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900004731.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004731.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004731.mm.1 |
TC1900000527.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004733.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004733.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004733.mm.1 |
TC1900000527.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004735.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004735.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004735.mm.1 |
TC1900000527.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004736.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004736.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004736.mm.1 |
TC1900000527.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004730.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004730.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004730.mm.1 |
TC1900000527.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004732.mm.1 |
NM_027314.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004732.mm.1 |
NM_027314.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004732.mm.1 |
TC1900000527.mm.1 |
MTA TC |