| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900003142.mm.1 |
NM_008137.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003142.mm.1 |
NM_008137.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003142.mm.1 |
TC1900000309.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003143.mm.1 |
NM_008137.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003143.mm.1 |
NM_008137.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003143.mm.1 |
TC1900000309.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003140.mm.1 |
NM_008137.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003140.mm.1 |
NM_008137.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003140.mm.1 |
TC1900000309.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003141.mm.1 |
NM_008137.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003141.mm.1 |
NM_008137.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003141.mm.1 |
TC1900000309.mm.1 |
MTA TC |