Assay Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198985.1 | NP_001185914.1 | 3 - 4 | 366 | 62 | |
| NM_011597.4 | NP_035727.2 | 3 - 4 | 356 | 62 | ||
| GenBank mRNA | AK028494.1 | - | 3 - 4 | 285 | 62 | |
| AK076689.1 | - | 3 - 4 | 438 | 62 | ||
| BC034677.1 | - | 3 - 4 | 3155078 | 62 | ||
| BC039924.1 | - | 3 - 4 | 3155079 | 62 | ||
| BY032291.1 | - | 3 - 4 | 358 | 62 | ||
| CB724174.1 | - | 3 - 4 | 366 | 62 | ||
More Information
Set Membership: |
|

Tjp2
Genomic Map