Assay Details
| Gene Symbol: Ap1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007457.2 | NP_031483.1 | 1 - 2 | 49 | 53 | |
| GenBank mRNA | AK002225.1 | - | 1 - 2 | 27 | 53 | |
| BC052692.1 | - | 1 - 2 | 53 | 5714625 | 53 | |
| M62418.1 | - | 1 - 2 | 49 | 53 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002225.1, BC052692.1, M62418.1, NM_007457.2 transcripts.
Set Membership: |
|

Ap1s1
Genomic Map