Assay Details
| Gene Symbol: Serpine1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008871.2 | NP_032897.2 | 2 - 3 | 417 | 65 | |
| GenBank mRNA | BC054091.1 | - | 2 - 3 | 414 | 30021925 | 65 |
| GQ872458.1 | - | 1 - 2 | 272 | 65 | ||
| GQ872459.1 | - | 1 - 2 | 272 | 65 | ||
| M33960.1 | - | 2 - 3 | 403 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpine1
Genomic Map