Assay Details
| Gene Symbol: Serpine1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008871.2 | NP_032897.2 | 3 - 4 | 654 | 79 | |
| GenBank mRNA | GQ872458.1 | - | 2 - 3 | 509 | 79 | |
More Information
Set Membership: |
|

Serpine1
Genomic Map