Assay Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018759.2 | NP_061229.2 | 2 - 3 | 175 | 78 | |
| NR_030777.1 | 2 - 3 | 175 | 78 | |||
| GenBank mRNA | AB012725.1 | - | 2 - 3 | 141 | 78 | |
| AK017693.1 | - | 2 - 3 | 176 | 78 | ||
| AK032801.1 | - | 2 - 3 | 176 | 78 | ||
| AK146438.1 | - | 2 - 3 | 156 | 78 | ||
| AV503563.1 | - | 2 - 3 | 186 | 78 | ||
| BC005567.1 | - | 2 - 3 | 166 | 78 | ||
| BC037055.1 | - | 2 - 3 | 180 | 4948371 | 78 | |
| BC039041.1 | - | 2 - 3 | 175 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC037055.1 transcripts.
Set Membership: |
|

Zfp326
Genomic Map