Assay Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018759.2 | NP_061229.2 | 3 - 4 | 208 | 77 | |
| GenBank mRNA | AB012725.1 | - | 3 - 4 | 174 | 77 | |
| AK017693.1 | - | 3 - 4 | 209 | 77 | ||
| AK146438.1 | - | 3 - 4 | 189 | 77 | ||
| AV503563.1 | - | 3 - 4 | 219 | 77 | ||
| BC005567.1 | - | 3 - 4 | 199 | 77 | ||
| BC039041.1 | - | 3 - 4 | 208 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp326
Genomic Map