Assay Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018759.2 | NP_061229.2 | 7 - 8 | 1035 | 138 | |
| NR_030777.1 | 5 - 6 | 571 | 138 | |||
| GenBank mRNA | AB012725.1 | - | 7 - 8 | 1001 | 138 | |
| AK017693.1 | - | 7 - 8 | 1036 | 138 | ||
| BC037055.1 | - | 5 - 6 | 576 | 4948371 | 138 | |
| BC039041.1 | - | 6 - 7 | 3040 | 138 | ||
| BU503985.1 | - | 4 - 5 | 419 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp326
Genomic Map