Assay Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018759.2 | NP_061229.2 | 8 - 9 | 1182 | 133 | |
| NR_030777.1 | 6 - 7 | 718 | 133 | |||
| GenBank mRNA | AB012725.1 | - | 8 - 9 | 1148 | 133 | |
| AK017693.1 | - | 8 - 9 | 1183 | 133 | ||
| AK084756.1 | - | 1 - 2 | 1142 | 133 | ||
| AK162729.1 | - | 2 - 3 | 186 | 133 | ||
| BC037055.1 | - | 6 - 7 | 723 | 4948371 | 133 | |
| BC039041.1 | - | 7 - 8 | 3187 | 133 | ||
| BU503985.1 | - | 5 - 6 | 566 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp326
Genomic Map