Assay Details
| Gene Symbol: Ssh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198109.4 | NP_932777.2 | 7 - 8 | 555 | 63 | |
| GenBank mRNA | AB099287.1 | - | 7 - 8 | 590 | 63 | |
| AK089308.1 | - | 7 - 8 | 559 | 63 | ||
| AK155557.1 | - | 7 - 8 | 580 | 63 | ||
| AK165380.1 | - | 6 - 7 | 520 | 63 | ||
| AK171210.1 | - | 7 - 8 | 604 | 63 | ||
| AK171556.1 | - | 7 - 8 | 570 | 63 | ||
| BC046529.1 | - | 1 - 2 | 37 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssh1
Genomic Map