Assay Details
| Gene Symbol: Gstm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010360.2 | NP_034490.1 | 3 - 4 | 290 | 85 | |
| GenBank mRNA | AK002750.1 | - | 3 - 4 | 222 | 85 | |
| AK019317.1 | - | 3 - 4 | 221 | 85 | ||
| AK028098.1 | - | 3 - 4 | 306 | 85 | ||
| BC008206.1 | - | 3 - 4 | 244 | 3154170 | 85 | |
| U24428.1 | - | 3 - 4 | 286 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gstm5
Genomic Map