Assay Details
| Gene Symbol: 2010012O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025563.3 | NP_079839.1 | 2 - 3 | 241 | 93 | |
| GenBank mRNA | AK007626.1 | - | 2 - 3 | 228 | 93 | |
| AK008215.1 | - | 2 - 3 | 236 | 93 | ||
| AK046524.1 | - | 2 - 3 | 241 | 93 | ||
| BC027506.1 | - | 2 - 3 | 213 | 1514880 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

2010012O05Rik
Genomic Map