Assay Details
| Gene Symbol: Cyp17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007809.3 | NP_031835.3 | 3 - 4 | 815 | 154 | |
| GenBank mRNA | AK132830.1 | - | 3 - 4 | 709 | 154 | |
| AK134376.1 | - | 3 - 4 | 709 | 154 | ||
| BC064793.1 | - | 3 - 4 | 789 | 30436278 | 154 | |
| M64863.1 | - | 3 - 4 | 688 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp17a1
Genomic Map