Assay Details
| Gene Symbol: Cyp17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007809.3 | NP_031835.3 | 7 - 8 | 1400 | 63 | |
| GenBank mRNA | AK132830.1 | - | 7 - 8 | 1294 | 63 | |
| AK134376.1 | - | 7 - 8 | 1294 | 63 | ||
| BC064793.1 | - | 7 - 8 | 1374 | 30436278 | 63 | |
| M64863.1 | - | 7 - 8 | 1273 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
glucocorticoid biosynthetic process
steroid metabolic process
hormone biosynthetic process
progesterone metabolic process
oxidation-reduction process
positive regulation of steroid hormone biosynthetic process
monooxygenase activity
steroid 17-alpha-monooxygenase activity
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
lyase activity
heme binding
metal ion binding
17-alpha-hydroxyprogesterone aldolase activity
steroid biosynthetic process
glucocorticoid biosynthetic process
steroid metabolic process
hormone biosynthetic process
progesterone metabolic process
oxidation-reduction process
positive regulation of steroid hormone biosynthetic process
monooxygenase activity
steroid 17-alpha-monooxygenase activity
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
lyase activity
heme binding
metal ion binding
17-alpha-hydroxyprogesterone aldolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | M64863.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | AK132830.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | AK134376.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | BC064793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | NM_007809.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | M64863.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | AK132830.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | AK134376.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | BC064793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P116813 | NM_007809.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23881 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155820 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | M64863.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | AK132830.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | AK134376.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | BC064793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | NM_007809.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | M64863.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | AK132830.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | AK134376.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | BC064793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P116813 | NM_007809.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | M64863.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | AK132830.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | AK134376.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | BC064793.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | NM_007809.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | M64863.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | AK132830.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | AK134376.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | BC064793.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116813 | NM_007809.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145317 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155820 | - | - |
| Whole Mouse Genome G4122A | A_51_P116813 | M64863.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116813 | AK132830.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116813 | AK134376.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116813 | BC064793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116813 | NM_007809.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116813 | M64863.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116813 | AK132830.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116813 | AK134376.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116813 | BC064793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116813 | NM_007809.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132464 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138605 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155820 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14306183 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14306182 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14498702 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14306180 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14306178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14357672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14306180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14387081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14387084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14257371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14182421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14357663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14387082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14187413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14387086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14306179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14352729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145331 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10468243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468241 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468247 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468248 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468245 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468240 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468246 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417017_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468239 | - | - |
| Mouse Genome 430 2.0 Array | 1417017_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417017_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13074_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13074_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa10480_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900014299.mm.1 | NM_007809.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014299.mm.1 | NM_007809.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014299.mm.1 | TC1900001612.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417017_PM_at | - | - |
| Mouse Expression Set 430 | 1417017_at | - | - |
| Murine Genome U74v2 Set | 102416_at | - | - |
| Murine Genome U74v2 Set | 102416_PM_at | - | - |
| Murine Genome U74v2 Set | 161721_PM_f_at | - | - |
| Murine Genome U74v2 Set | 161721_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417017_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417017_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417017_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417017_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 630817 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1340408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340408 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2648251 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2648251 | - | - |
| mouse-6 v1.1 (Illumina) | scl52399.8.1_189-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340408 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52399.8.1_189-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340408 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2874352 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2874352 | - | - |

Cyp17a1
Genomic Map