Assay Details
| Gene Symbol: Nt5c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164363.1 | NP_001157835.1 | 15 - 16 | 1720 | 99 | |
| NM_001164365.1 | NP_001157837.1 | 15 - 16 | 1377 | 99 | ||
| NM_029810.4 | NP_084086.3 | 15 - 16 | 1308 | 99 | ||
| NR_028353.1 | 16 - 17 | 1367 | 99 | |||
| GenBank mRNA | AK008045.1 | - | 15 - 16 | 1255 | 99 | |
| AK030786.1 | - | 16 - 17 | 1327 | 99 | ||
| AK036730.1 | - | 15 - 16 | 1266 | 99 | ||
| AK038458.1 | - | 12 - 13 | 1014 | 99 | ||
| AK132384.1 | - | 15 - 16 | 1266 | 99 | ||
| AK147901.1 | - | 15 - 16 | 1266 | 99 | ||
| AK162385.1 | - | 15 - 16 | 1685 | 99 | ||
| AK169616.1 | - | 15 - 16 | 1266 | 99 | ||
| BC064760.1 | - | 14 - 15 | 1242 | 30436992 | 99 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC006028.1 | - | Chr. - - [-] on Build | 99 | |||
More Information
Set Membership: |
|

Nt5c2
Genomic Map