Assay Details
| Gene Symbol: Nt5c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164363.1 | NP_001157835.1 | 3 - 4 | 683 | 105 | |
| NM_001164365.1 | NP_001157837.1 | 3 - 4 | 340 | 105 | ||
| NM_029810.4 | NP_084086.3 | 3 - 4 | 271 | 105 | ||
| GenBank mRNA | AK008045.1 | - | 3 - 4 | 218 | 105 | |
| AK036730.1 | - | 3 - 4 | 229 | 105 | ||
| AK132384.1 | - | 3 - 4 | 229 | 105 | ||
| AK147901.1 | - | 3 - 4 | 229 | 105 | ||
| AK169616.1 | - | 3 - 4 | 229 | 105 | ||
| AK201286.1 | - | 1 - 2 | 73 | 105 | ||
| AK211414.1 | - | 1 - 2 | 73 | 105 | ||
| BC064760.1 | - | 2 - 3 | 205 | 30436992 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c2
Genomic Map