Assay Details
| Gene Symbol: Nt5c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164363.1 | NP_001157835.1 | 5 - 6 | 903 | 89 | |
| NM_001164365.1 | NP_001157837.1 | 5 - 6 | 560 | 89 | ||
| NM_029810.4 | NP_084086.3 | 5 - 6 | 491 | 89 | ||
| NR_028353.1 | 6 - 7 | 550 | 89 | |||
| GenBank mRNA | AK008045.1 | - | 5 - 6 | 438 | 89 | |
| AK030786.1 | - | 6 - 7 | 508 | 89 | ||
| AK036730.1 | - | 5 - 6 | 449 | 89 | ||
| AK038458.1 | - | 2 - 3 | 196 | 89 | ||
| AK132384.1 | - | 5 - 6 | 449 | 89 | ||
| AK147901.1 | - | 5 - 6 | 449 | 89 | ||
| AK162385.1 | - | 5 - 6 | 869 | 89 | ||
| AK169616.1 | - | 5 - 6 | 449 | 89 | ||
| AK181552.1 | - | 4 - 5 | 352 | 89 | ||
| AK187621.1 | - | 4 - 5 | 352 | 89 | ||
| BC064760.1 | - | 4 - 5 | 425 | 30436992 | 89 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC006028.1 | - | Chr. - - [-] on Build | 89 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nt5c2
Genomic Map