Assay Details
| Gene Symbol: Cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033569.3 | NP_291047.2 | 6 - 7 | 2392 | 58 | |
| GenBank mRNA | AF216961.1 | - | 6 - 7 | 1704 | 58 | |
| AK032006.1 | - | 6 - 7 | 2351 | 58 | ||
| AK053711.1 | - | 6 - 7 | 2410 | 58 | ||
| BC038027.1 | - | 284 | 58 | |||
| BC052513.1 | - | 4 - 5 | 407 | 58 | ||
| BC125033.1 | - | 6 - 7 | 1704 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnnm2
Genomic Map