Assay Details
| Gene Symbol: Tsks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011651.2 | NP_035781.2 | 10 - 11 | 1643 | 78 | |
| GenBank mRNA | AF025310.2 | - | 10 - 11 | 1622 | 78 | |
| BX630702.1 | - | 1 - 2 | 78 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259631 | BX630702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259631 | NM_011651.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259631 | AF025310.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259631 | NM_001077591.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259631 | BX630702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259631 | NM_011651.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259631 | AF025310.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P259631 | NM_001077591.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552420 | AB021968.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552420 | AF164524.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552420 | AF186188.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552420 | AK004045.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552420 | AK015095.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552420 | BC089577.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552420 | NM_019775.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330797 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618255 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91671 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122261 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259631 | BX630702.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259631 | NM_011651.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259631 | AF025310.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259631 | NM_001077591.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259631 | BX630702.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259631 | NM_011651.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259631 | AF025310.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P259631 | NM_001077591.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090256 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06181436 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122261 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01506881 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101328 | - | - |
| Whole Mouse Genome G4122A | A_51_P259631 | BX630702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259631 | NM_011651.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259631 | AF025310.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259631 | NM_001077591.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259631 | BX630702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259631 | NM_011651.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259631 | AF025310.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P259631 | NM_001077591.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127578 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130016 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122261 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101328 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06143479 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06143482 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06143483 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06143481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06359033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06662564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06631240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05938480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06395745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06574444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06143481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06024160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06359028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06492751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05998133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06569969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06561169 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552803 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552801 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552809 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552799 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552807 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552805 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552808 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552804 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552806 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552802 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419439_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552796 | - | - |
| Mouse Genome 430 2.0 Array | 1419439_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419439_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22116_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22116_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700006977.mm.1 | NM_001077591.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006977.mm.1 | NM_001077591.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006977.mm.1 | NM_011651.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006977.mm.1 | NM_011651.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006977.mm.1 | TC0700000822.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006978.mm.1 | NM_001077591.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006978.mm.1 | NM_001077591.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006978.mm.1 | NM_011651.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006978.mm.1 | NM_011651.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006978.mm.1 | TC0700000822.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419439_at | - | - |
| Mouse Expression Set 430 | 1419439_PM_at | - | - |
| Murine Genome U74v2 Set | 94624_PM_at | - | - |
| Murine Genome U74v2 Set | 94624_at | - | - |
| Murine Genome U74v2 Set | 170429_i_at | - | - |
| Murine Genome U74v2 Set | 170429_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419439_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419439_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419439_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419439_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 857718 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4120022 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120022 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235441 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235441 | - | - |
| mouse-6 v1.1 (Illumina) | scl32700.5.1_108-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120022 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32700.5.1_108-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120022 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3138175 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3138175 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235441 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32700.5.1_108-S | - | - |

Tsks
Genomic Map