Assay Details
| Gene Symbol: Tsks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077591.1 | NP_001071059.1 | 1 - 2 | 209 | 78 | |
| NM_011651.2 | NP_035781.2 | 1 - 2 | 209 | 78 | ||
| GenBank mRNA | AF025310.2 | - | 1 - 2 | 189 | 78 | |
| BC118618.1 | - | 1 - 2 | 188 | 40107271 | 78 | |
| BC121827.1 | - | 1 - 2 | 188 | 40107270 | 78 | |
| CJ047818.1 | - | 1 - 2 | 210 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tsks
Genomic Map