Assay Details
| Gene Symbol: Tsks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077591.1 | NP_001071059.1 | 9 - 10 | 1497 | 70 | |
| NM_011651.2 | NP_035781.2 | 9 - 10 | 1524 | 70 | ||
| GenBank mRNA | AF025310.2 | - | 9 - 10 | 1504 | 70 | |
| BC118618.1 | - | 9 - 10 | 1476 | 40107271 | 70 | |
| BC121827.1 | - | 9 - 10 | 1476 | 40107270 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsks
Genomic Map