Assay Details
| Gene Symbol: Aldh16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145954.1 | NP_666066.1 | 13 - 14 | 1800 | 66 | |
| GenBank mRNA | AK030673.1 | - | 13 - 14 | 1793 | 66 | |
| AK144967.1 | - | 1799 | 66 | |||
| AK144995.1 | - | 13 - 14 | 1807 | 66 | ||
| AK151691.1 | - | 13 - 14 | 1800 | 66 | ||
| AK153129.1 | - | 6 - 7 | 707 | 66 | ||
| AK161198.1 | - | 13 - 14 | 1838 | 66 | ||
| AK164374.1 | - | 13 - 14 | 1846 | 66 | ||
| AK169346.1 | - | 2151 | 66 | |||
| AK170276.1 | - | 13 - 14 | 1839 | 66 | ||
| AK212056.1 | - | 2 - 3 | 243 | 66 | ||
| AK219145.1 | - | 3 - 4 | 353 | 66 | ||
| AK220200.1 | - | 13 - 14 | 1776 | 66 | ||
| BC013548.1 | - | 13 - 14 | 1800 | 4222058 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh16a1
Genomic Map