Assay Details
| Gene Symbol: Aldh16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145954.1 | NP_666066.1 | 10 - 11 | 1386 | 86 | |
| GenBank mRNA | AK030673.1 | - | 10 - 11 | 1379 | 86 | |
| AK144967.1 | - | 1385 | 86 | |||
| AK144995.1 | - | 10 - 11 | 1393 | 86 | ||
| AK151691.1 | - | 10 - 11 | 1386 | 86 | ||
| AK153129.1 | - | 3 - 4 | 293 | 86 | ||
| AK161198.1 | - | 10 - 11 | 1424 | 86 | ||
| AK164374.1 | - | 10 - 11 | 1432 | 86 | ||
| AK169346.1 | - | 1737 | 86 | |||
| AK170276.1 | - | 10 - 11 | 1425 | 86 | ||
| AK220200.1 | - | 10 - 11 | 1362 | 86 | ||
| BC013548.1 | - | 10 - 11 | 1386 | 4222058 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh16a1
Genomic Map