Assay Details
| Gene Symbol: Bax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007527.3 | NP_031553.1 | 2 - 3 | 216 | 85 | |
| GenBank mRNA | AK013298.1 | - | 2 - 3 | 126 | 85 | |
| AK149994.1 | - | 2 - 3 | 109 | 85 | ||
| AK159233.1 | - | 2 - 3 | 181 | 85 | ||
| AY095934.1 | - | 1 - 2 | 563 | 85 | ||
| BC018228.1 | - | 2 - 3 | 161 | 3968903 | 85 | |
| BC053380.1 | - | 2 - 3 | 133 | 6489475 | 85 | |
| BY308683.1 | - | 2 - 3 | 196 | 85 | ||
| CJ078904.1 | - | 2 - 3 | 217 | 85 | ||
| L22472.1 | - | 2 - 3 | 93 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY095934.1 transcripts.
Set Membership: |
|

Bax
Genomic Map