Assay Details
| Gene Symbol: Ppp1r16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033371.2 | NP_203535.1 | 4 - 5 | 1281 | 62 | |
| GenBank mRNA | AK150235.1 | - | 1 - 2 | 75 | 62 | |
| AK154865.1 | - | 4 - 5 | 1266 | 62 | ||
| AK171452.1 | - | 4 - 5 | 1266 | 62 | ||
| AK220248.1 | - | 4 - 5 | 1281 | 62 | ||
| AY010723.1 | - | 3 - 4 | 476 | 62 | ||
| BC033318.1 | - | 3 - 4 | 841 | 62 | ||
| BC044775.1 | - | 3 - 4 | 963 | 62 | ||
| BC049958.1 | - | 3 - 4 | 963 | 62 | ||
| BC057450.1 | - | 3 - 4 | 898 | 3966334 | 62 | |
| BC096620.1 | - | 4 - 5 | 1281 | 6390076 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r16a
Genomic Map