Assay Details
| Gene Symbol: Ppp1r16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033371.2 | NP_203535.1 | 6 - 7 | 1512 | 70 | |
| GenBank mRNA | AK154865.1 | - | 6 - 7 | 1497 | 70 | |
| AK171452.1 | - | 6 - 7 | 1497 | 70 | ||
| AY010723.1 | - | 5 - 6 | 707 | 70 | ||
| BC044775.1 | - | 4 - 5 | 1267 | 70 | ||
| BC049958.1 | - | 4 - 5 | 1267 | 70 | ||
| BC057450.1 | - | 5 - 6 | 1129 | 3966334 | 70 | |
| BC096620.1 | - | 6 - 7 | 1512 | 6390076 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r16a
Genomic Map