Assay Details
| Gene Symbol: Recql4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058214.3 | NP_478121.2 | 10 - 11 | 1752 | 64 | |
| GenBank mRNA | AB039882.1 | - | 10 - 11 | 1625 | 64 | |
| AB175741.1 | - | 10 - 11 | 1669 | 64 | ||
| AK090260.1 | - | 11 - 12 | 1356 | 64 | ||
| BC064680.1 | - | 10 - 11 | 1726 | 64 | ||
| BC131656.1 | - | 10 - 11 | 1665 | 64 | ||
| BC131657.1 | - | 9 - 10 | 1852 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Recql4
Genomic Map