Assay Details
| Gene Symbol: Pgam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163538.1 | NP_001157010.1 | 1 - 2 | 250 | 83 | |
| NM_028273.3 | NP_082549.2 | 1 - 2 | 250 | 83 | ||
| GenBank mRNA | AK034588.1 | - | 1 - 2 | 234 | 83 | |
| AK146216.1 | - | 1 - 2 | 251 | 83 | ||
| AK162266.1 | - | 1 - 2 | 201 | 83 | ||
| AK169643.1 | - | 1 - 2 | 213 | 83 | ||
| BC021317.1 | - | 1 - 2 | 200 | 83 | ||
| BC052179.1 | - | 1 - 2 | 174 | 83 | ||
| BC138924.1 | - | 1 - 2 | 233 | 8861946 | 83 | |
| BC138925.1 | - | 1 - 2 | 233 | 8861947 | 83 | |
| BC145467.1 | - | 1 - 2 | 233 | 9054029 | 83 | |
| BY101410.1 | - | 1 - 2 | 264 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pgam5
Genomic Map