Assay Details
| Gene Symbol: Vsig10l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290316.1 | NP_001277245.1 | 2 - 3 | 553 | 67 | |
| GenBank mRNA | BC112410.1 | - | 2 - 3 | 547 | 67 | |
More Information
Set Membership: |
|

Vsig10l
Genomic Map