Assay Details
| Gene Symbol: Colec10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173422.3 | NP_775598.2 | 3 - 4 | 300 | 63 | |
| GenBank mRNA | AB016429.1 | - | 3 - 4 | 304 | 63 | |
| AK028423.1 | - | 3 - 4 | 302 | 63 | ||
| BC106836.1 | - | 3 - 4 | 302 | 40044882 | 63 | |
| BC106837.1 | - | 3 - 4 | 302 | 40044883 | 63 | |
| BC108945.1 | - | 3 - 4 | 302 | 40043083 | 63 | |
More Information
Set Membership: |
|

Colec10
Genomic Map