Assay Details
| Gene Symbol: Nyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175521.3 | NP_780730.2 | 4 - 5 | 2417 | 87 | |
| GenBank mRNA | AB429289.1 | - | 4 - 5 | 2414 | 87 | |
| AK032567.1 | - | 2 - 3 | 1587 | 87 | ||
| AK080821.1 | - | 5 - 6 | 2784 | 87 | ||
| AK141276.1 | - | 5 - 6 | 2271 | 87 | ||
| AK148980.1 | - | 2 - 3 | 1378 | 87 | ||
| BC057372.1 | - | 4 - 5 | 2417 | 5686602 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nyap1
Genomic Map