Assay Details
| Gene Symbol: Gigyf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031408.2 | NP_113596.2 | 12 - 13 | 1537 | 80 | |
| GenBank mRNA | AK220289.1 | - | 5 - 6 | 546 | 80 | |
| AY176042.1 | - | 12 - 13 | 1553 | 80 | ||
| BC052875.1 | - | 6 - 7 | 641 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gigyf1
Genomic Map