Assay Details
| Gene Symbol: Gigyf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031408.2 | NP_113596.2 | 8 - 9 | 1017 | 100 | |
| GenBank mRNA | AY176042.1 | - | 8 - 9 | 1033 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gigyf1
Genomic Map