Assay Details
| Gene Symbol: Cyp2w1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160265.1 | NP_001153737.1 | 6 - 7 | 1000 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2w1| Gene Symbol: Cyp2w1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160265.1 | NP_001153737.1 | 6 - 7 | 1000 | 66 | |
Set Membership: |
|