Assay Details
| Gene Symbol: Slc25a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007450.4 | NP_031476.3 | 2 - 3 | 691 | 82 | |
| GenBank mRNA | AK078077.1 | - | 2 - 3 | 713 | 82 | |
| AK177376.1 | - | 1 - 2 | 173 | 82 | ||
| BC003791.1 | - | 2 - 3 | 691 | 3599576 | 82 | |
| BC026925.1 | - | 2 - 3 | 702 | 4500784 | 82 | |
| U27315.1 | - | 2 - 3 | 690 | 82 | ||
| X74510.1 | - | 2 - 3 | 697 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a4
Genomic Map