Assay Details
| Gene Symbol: Ndufb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023172.3 | NP_075661.1 | 3 - 4 | 443 | 95 | |
| GenBank mRNA | AK002254.1 | - | 3 - 4 | 444 | 95 | |
| AK003540.1 | - | 3 - 4 | 444 | 95 | ||
| AK004522.1 | - | 3 - 4 | 444 | 95 | ||
| BC031539.1 | - | 3 - 4 | 425 | 3979845 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30251032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30251033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30251034 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | BC031539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | AK004522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | NM_023172.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | AJ300739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | AK002254.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | BC031539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | AK004522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | NM_023172.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | AJ300739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177552 | AK002254.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262815 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22048 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113568 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | BC031539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | AK004522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | NM_023172.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | AJ300739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | AK002254.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | BC031539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | AK004522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | NM_023172.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | AJ300739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177552 | AK002254.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | BC031539.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | AK004522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | NM_023172.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | AJ300739.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | AK002254.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | BC031539.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | AK004522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | NM_023172.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | AJ300739.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177552 | AK002254.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30251054 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113568 | - | - |
| Whole Mouse Genome G4122A | A_51_P177552 | BC031539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177552 | AK004522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177552 | NM_023172.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177552 | AJ300739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177552 | AK002254.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177552 | BC031539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177552 | AK004522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177552 | NM_023172.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177552 | AJ300739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177552 | AK002254.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111397 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113568 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11981698 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11981697 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11981691 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11981701 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30251033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30251032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30251034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30251062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12172777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12063689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12172795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11861400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12172769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12172782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30251077 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10424343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424344 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452184_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455934_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436803_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10424340 | - | - |
| Mouse Genome 430 2.0 Array | 1455934_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436803_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455934_at | - | - |
| Mouse Genome 430 2.0 Array | 1452184_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452184_at | - | - |
| Mouse Genome 430 2.0 Array | 1436803_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66218_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66218_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500003006.mm.1 | NM_023172.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003006.mm.1 | NM_023172.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003006.mm.1 | TC1500000423.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455934_PM_at | - | - |
| Mouse Expression Set 430 | 1452184_PM_at | - | - |
| Mouse Expression Set 430 | 1455934_at | - | - |
| Mouse Expression Set 430 | 1436803_a_at | - | - |
| Mouse Expression Set 430 | 1452184_at | - | - |
| Mouse Expression Set 430 | 1436803_PM_a_at | - | - |
| Murine Genome U74v2 Set | 100079_at | - | - |
| Murine Genome U74v2 Set | 100079_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452184_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452184_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436803_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455934_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436803_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455934_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452184_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452184_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436803_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455934_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455934_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436803_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 909902 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3780348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3780348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2605004 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2605004 | - | - |
| mouse-6 v1.1 (Illumina) | scl066218.3_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066218.3_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2605004 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2605004 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066218.3_27-S | - | - |

Ndufb9
Genomic Map