Assay Details
| Gene Symbol: Vsig10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033311.3 | NP_001028483.2 | 1 - 2 | 241 | 119 | |
| GenBank mRNA | AK138152.1 | - | 1 - 2 | 190 | 119 | |
| CJ057085.1 | - | 1 - 2 | 190 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK138152.1, NM_001033311.3 transcripts.
Set Membership: |
|

Vsig10
Genomic Map