Assay Details
| Gene Symbol: Lrwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027891.4 | NP_082167.2 | 12 - 13 | 1658 | 80 | |
| GenBank mRNA | AK004717.1 | - | 10 - 11 | 2620 | 80 | |
| AK084948.1 | - | 12 - 13 | 1658 | 80 | ||
| AK085327.1 | - | 1 - 2 | 1689 | 80 | ||
| AK153976.1 | - | 12 - 13 | 1629 | 80 | ||
| AK154352.1 | - | 12 - 13 | 1600 | 80 | ||
| AK167020.1 | - | 12 - 13 | 1607 | 80 | ||
| BC048397.1 | - | 12 - 13 | 1637 | 6396815 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrwd1
Genomic Map