Assay Details
| Gene Symbol: Lrwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027891.4 | NP_082167.2 | 6 - 7 | 924 | 94 | |
| GenBank mRNA | AK004717.1 | - | 4 - 5 | 1886 | 94 | |
| AK042370.1 | - | 3 - 4 | 2174 | 94 | ||
| AK051910.1 | - | 6 - 7 | 872 | 94 | ||
| AK084948.1 | - | 6 - 7 | 924 | 94 | ||
| AK153976.1 | - | 6 - 7 | 895 | 94 | ||
| AK154352.1 | - | 6 - 7 | 866 | 94 | ||
| AK167020.1 | - | 6 - 7 | 873 | 94 | ||
| AK172708.1 | - | 6 - 7 | 892 | 94 | ||
| BC048397.1 | - | 6 - 7 | 903 | 6396815 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrwd1
Genomic Map