Assay Details
| Gene Symbol: Ddb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015735.1 | NP_056550.1 | 11 - 12 | 1384 | 79 | |
| GenBank mRNA | AB026432.1 | - | 11 - 12 | 1384 | 79 | |
| AF159853.1 | - | 11 - 12 | 1344 | 79 | ||
| AK146522.1 | - | 11 - 12 | 1442 | 79 | ||
| AK154303.1 | - | 11 - 12 | 1410 | 79 | ||
| AK155020.1 | - | 11 - 12 | 1465 | 79 | ||
| AK155920.1 | - | 11 - 12 | 1425 | 79 | ||
| AK157491.1 | - | 11 - 12 | 1464 | 79 | ||
| AK209080.1 | - | 1 - 2 | 53 | 79 | ||
| BC009661.1 | - | 11 - 12 | 1391 | 3487617 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddb1
Genomic Map