Assay Details
| Gene Symbol: Cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037801.2 | NP_001032890.1 | 7 - 8 | 1486 | 85 | |
| NM_009852.3 | NP_033982.3 | 7 - 8 | 1486 | 85 | ||
| GenBank mRNA | AK030822.1 | - | 7 - 8 | 1456 | 85 | |
| AK030823.1 | - | 7 - 8 | 1487 | 85 | ||
| AK088050.1 | - | 7 - 8 | 1464 | 85 | ||
| AK141213.1 | - | 3 - 4 | 633 | 85 | ||
| BC014294.1 | - | 7 - 8 | 1492 | 4187689 | 85 | |
| U12434.1 | - | 7 - 8 | 1284 | 85 | ||
| U35370.1 | - | 7 - 8 | 1402 | 85 | ||
| U37543.1 | - | 7 - 8 | 1443 | 85 | ||
| U37544.1 | - | 7 - 8 | 1443 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd6
Genomic Map