Assay Details
| Gene Symbol: Prpf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253843.1 | NP_001240772.1 | 8 - 9 | 919 | 82 | |
| NM_001253844.1 | NP_001240773.1 | 6 - 7 | 512 | 82 | ||
| NM_134129.4 | NP_598890.1 | 8 - 9 | 862 | 82 | ||
| GenBank mRNA | AF251503.1 | - | 8 - 9 | 827 | 82 | |
| AK047693.1 | - | 8 - 9 | 737 | 82 | ||
| AK076998.1 | - | 6 - 7 | 513 | 82 | ||
| AK088769.1 | - | 8 - 9 | 847 | 82 | ||
| AK164681.1 | - | 8 - 9 | 850 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prpf19
Genomic Map