Assay Details
| Gene Symbol: Cyb561a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282065.1 | NP_001268994.1 | 6 - 7 | 1750 | 58 | |
| GenBank mRNA | AK160179.1 | - | 6 - 7 | 1454 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyb561a3
Genomic Map