Assay Details
| Gene Symbol: Lrrc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019457.2 | NP_062330.1 | 2 - 3 | 306 | 148 | |
| GenBank mRNA | AF092208.1 | - | 2 - 3 | 251 | 148 | |
| AK016236.1 | - | 2 - 3 | 295 | 148 | ||
| AK077124.1 | - | 2 - 3 | 290 | 148 | ||
| BC046277.1 | - | 2 - 3 | 257 | 6310345 | 148 | |
| CJ046721.1 | - | 2 - 3 | 307 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc6
Genomic Map